Search
Search

STRADA| Gene Symbol: STRADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116251.4 | XP_001116251.1 | 674 | 89 | ||
| XM_001116259.4 | XP_001116259.1 | 724 | 89 | |||
| XM_001116272.4 | XP_001116272.1 | 764 | 89 | |||
| XM_001116286.4 | XP_001116286.1 | 639 | 89 | |||
| XM_015120084.2 | XP_014975570.1 | 741 | 89 | |||
| XM_015120085.2 | XP_014975571.1 | 727 | 89 | |||
| XM_015120088.2 | XP_014975574.1 | 764 | 89 | |||
| XM_015120089.2 | XP_014975575.1 | 666 | 89 | |||
| XM_015120090.2 | XP_014975576.1 | 712 | 89 | |||
| XM_028836646.1 | XP_028692479.1 | 654 | 89 | |||
| XM_028836647.1 | XP_028692480.1 | 666 | 89 | |||
| XM_028836648.1 | XP_028692481.1 | 660 | 89 | |||
| XM_028836649.1 | XP_028692482.1 | 615 | 89 | |||
| GenBank mRNA | JU321857.1 | - | 687 | 89 | ||
| JU321978.1 | - | 716 | 89 | |||
| JU337100.1 | - | 629 | 89 | |||
Set Membership: |
|