Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02831565_m1
See other NUMB GE Assays ›
Gene Symbol:
NUMB
Gene Name:
NUMB endocytic adaptor protein
Gene Aliases:
-
Chromosome Location:
Chr.7: 135402244 - 135596342 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02831565_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: NUMB
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015144051.2
XP_014999537.2
482
107
XM_015144052.2
XP_014999538.2
322
107
XM_015144053.2
XP_014999539.2
410
107
XM_015144054.2
XP_014999540.2
482
107
XM_015144056.2
XP_014999542.2
482
107
XM_015144057.2
XP_014999543.2
320
107
XM_015144058.2
XP_014999544.2
410
107
XM_015144059.2
XP_014999545.2
482
107
XM_015144060.2
XP_014999546.2
410
107
XM_015144061.2
XP_014999547.2
482
107
XM_015144062.2
XP_014999548.2
410
107
XM_015144063.2
XP_014999549.2
482
107
XM_015144064.2
XP_014999550.2
482
107
XM_028851650.1
XP_028707483.1
257
107
XM_028851651.1
XP_028707484.1
165
107
XM_028851654.1
XP_028707487.1
327
107
XM_028851655.1
XP_028707488.1
410
107
XM_028851656.1
XP_028707489.1
482
107
GenBank mRNA
JU322008.1
-
417
107
JU322039.1
-
417
107
JU336884.1
-
417
107
JU472303.1
-
443
107
JU477624.1
-
443
107
JU477625.1
-
443
107
JV044154.1
-
440
107
JV044155.1
-
440
107
JV048553.1
-
440
107
JV048554.1
-
440
107
JV490407.1
-
417
107
JV636433.1
-
423
107
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Macaca mulatta
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline