Search
Search

SLC26A10| Gene Symbol: SLC26A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152123.2 | XP_015007609.1 | 1136 | 78 | ||
| XM_028829667.1 | XP_028685500.1 | 1137 | 78 | |||
| XM_028829668.1 | XP_028685501.1 | 1138 | 78 | |||
| XM_028829669.1 | XP_028685502.1 | 1139 | 78 | |||
| XM_028829670.1 | XP_028685503.1 | 1139 | 78 | |||
Set Membership: |
|