Search
Search

SPAG8| Gene Symbol: SPAG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117473.2 | XP_014972959.1 | 1178 | 87 | ||
| XM_015117475.2 | XP_014972961.1 | 1179 | 87 | |||
| XM_015117478.2 | XP_014972964.1 | 1178 | 87 | |||
| XM_028835076.1 | XP_028690909.1 | 1179 | 87 | |||
| XM_028835077.1 | XP_028690910.1 | 1179 | 87 | |||
Set Membership: |
|