Search
Search

EMP2| Gene Symbol: EMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265948.1 | NP_001252877.1 | 141 | |||
| XM_015125595.2 | XP_014981081.2 | 283 | 141 | |||
| GenBank mRNA | JU333314.1 | - | 141 | |||
| JV046402.1 | - | 141 | ||||
| JV709058.1 | - | 223 | 141 | |||
Set Membership: |
|