Search
Search

NR6A1| Gene Symbol: NR6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834859.1 | XP_028690692.1 | 598 | 65 | ||
| XM_028834860.1 | XP_028690693.1 | 598 | 65 | |||
| XM_028834861.1 | XP_028690694.1 | 1746 | 65 | |||
| GenBank mRNA | JV635858.1 | - | 561 | 65 | ||
Set Membership: |
|