Search
Search

ACP5| Gene Symbol: ACP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258183.1 | NP_001245112.1 | 645 | 162 | ||
| XM_015122923.2 | XP_014978409.1 | 710 | 162 | |||
| XM_015122924.2 | XP_014978410.1 | 638 | 162 | |||
| GenBank mRNA | JU476047.1 | - | 645 | 162 | ||
Set Membership: |
|