Search
Search

CAV1| Gene Symbol: CAV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168614.1 | NP_001162085.1 | 251 | 66 | ||
| XM_015134736.2 | XP_014990222.1 | 874 | 66 | |||
| XM_028844899.1 | XP_028700732.1 | 263 | 66 | |||
| GenBank mRNA | CO579359.1 | - | 251 | 66 | ||
| JU336862.1 | - | 270 | 66 | |||
Set Membership: |
|