Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257434.1 | NP_001244363.1 | 231 | 103 | ||
GenBank mRNA | JU335861.1 | - | 231 | 103 | ||
JU476734.1 | - | 229 | 103 | |||
JV043626.1 | - | 227 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |