Search
Search

ST3GAL1| Gene Symbol: ST3GAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261634.1 | NP_001248563.1 | 1089 | 89 | ||
| XM_015146017.2 | XP_015001503.1 | 1654 | 89 | |||
| XM_015146018.2 | XP_015001504.1 | 1797 | 89 | |||
| XM_015146019.2 | XP_015001505.1 | 2220 | 89 | |||
| XM_028852132.1 | XP_028707965.1 | 2077 | 89 | |||
| XM_028852133.1 | XP_028707966.1 | 2073 | 89 | |||
| XM_028852134.1 | XP_028707967.1 | 1945 | 89 | |||
| XM_028852135.1 | XP_028707968.1 | 1998 | 89 | |||
| XM_028852136.1 | XP_028707969.1 | 1957 | 89 | |||
| GenBank mRNA | JU473118.1 | - | 1292 | 89 | ||
| JU473119.1 | - | 1089 | 89 | |||
Set Membership: |
|