Search
Search

NFE2L1| Gene Symbol: NFE2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261099.2 | NP_001248028.1 | 1595 | 67 | ||
| XM_015119374.2 | XP_014974860.1 | 1662 | 67 | |||
| XM_015119377.2 | XP_014974863.1 | 954 | 67 | |||
| XM_015119378.2 | XP_014974864.1 | 810 | 67 | |||
| XM_028835475.1 | XP_028691308.1 | 1559 | 67 | |||
| GenBank mRNA | JU320594.1 | - | 1597 | 67 | ||
| JV043404.1 | - | 1596 | 67 | |||
| JV636580.1 | - | 1532 | 67 | |||
Set Membership: |
|