Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: CST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098076.4 | XP_001098076.1 | 316 | 146 | ||
GenBank mRNA | JU477569.1 | - | 175 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |