Search
Search

PSMG1| Gene Symbol: PSMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109110.4 | XP_001109110.2 | 910 | 87 | ||
| XM_028845322.1 | XP_028701155.1 | 1104 | 87 | |||
| XM_028845323.1 | XP_028701156.1 | 784 | 87 | |||
| GenBank mRNA | JU322893.1 | - | 804 | 87 | ||
| JU495446.1 | - | 879 | 87 | |||
| JV045763.1 | - | 865 | 87 | |||
Set Membership: |
|