Search
Search

KLF4| Gene Symbol: KLF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142793.2 | NP_001136265.1 | 1402 | 127 | ||
| GenBank mRNA | FJ375752.1 | - | 1092 | 127 | ||
| JV727062.1 | - | 199 | 127 | |||
Set Membership: |
|