Search
Search

BCL9| Gene Symbol: BCL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261320.1 | NP_001248249.1 | 1197 | 75 | ||
| XM_015149484.2 | XP_015004970.1 | 1223 | 75 | |||
| XM_015149493.2 | XP_015004979.1 | 1371 | 75 | |||
| XM_015149498.2 | XP_015004984.1 | 1149 | 75 | |||
| XM_028835532.1 | XP_028691365.1 | 1052 | 75 | |||
| GenBank mRNA | JU321966.1 | - | 1203 | 75 | ||
| JU476687.1 | - | 1260 | 75 | |||
| JV047863.1 | - | 1197 | 75 | |||