Search
Search

RERG| Gene Symbol: RERG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261674.1 | NP_001248603.1 | 238 | 146 | ||
| XM_015151102.2 | XP_015006588.1 | 602 | 146 | |||
| GenBank mRNA | JU319806.1 | - | 238 | 146 | ||
Set Membership: |
|