Search
Search

PTPRU| Gene Symbol: PTPRU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133466.2 | XP_014988952.2 | 583 | 68 | ||
| XM_015133475.2 | XP_014988961.2 | 583 | 68 | |||
| XM_015133484.2 | XP_014988970.2 | 583 | 68 | |||
| XM_015133492.2 | XP_014988978.2 | 583 | 68 | |||
| XM_015133502.2 | XP_014988988.1 | 583 | 68 | |||
| XM_015133509.2 | XP_014988995.1 | 583 | 68 | |||
| XM_015133513.2 | XP_014988999.1 | 598 | 68 | |||
| XM_015133520.2 | XP_014989006.1 | 583 | 68 | |||
| XM_015133528.2 | XP_014989014.1 | 583 | 68 | |||
| XM_015133536.2 | XP_014989022.1 | 593 | 68 | |||
| XM_015133544.2 | XP_014989030.1 | 598 | 68 | |||
| XM_028837043.1 | XP_028692876.1 | 583 | 68 | |||
| XM_028837061.1 | XP_028692894.1 | 444 | 68 | |||
| GenBank mRNA | JV602886.1 | - | 581 | 68 | ||
Set Membership: |
|