Search
Search

SLC27A4| Gene Symbol: SLC27A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116713.2 | XP_014972199.1 | 1900 | 96 | ||
| XM_015116714.2 | XP_014972200.1 | 1893 | 96 | |||
| XM_015116719.2 | XP_014972205.1 | 1965 | 96 | |||
| XM_028834768.1 | XP_028690601.1 | 2142 | 96 | |||
| GenBank mRNA | JU322525.1 | - | 1781 | 96 | ||
| JV045501.1 | - | 1631 | 96 | |||
Set Membership: |
|