Search
Search

TROAP| Gene Symbol: TROAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109203.4 | XP_001109203.1 | 360 | 88 | ||
| XM_015113941.2 | XP_014969427.1 | 355 | 88 | |||
| XM_015113942.2 | XP_014969428.1 | 360 | 88 | |||
| XM_028829449.1 | XP_028685282.1 | 357 | 88 | |||
| GenBank mRNA | JU478430.1 | - | 179 | 88 | ||
| JV639588.1 | - | 183 | 88 | |||
Set Membership: |
|