Search
Search

KARS1| Gene Symbol: KARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257593.1 | NP_001244522.1 | 1477 | 81 | ||
| XM_015126647.2 | XP_014982133.2 | 1419 | 81 | |||
| GenBank mRNA | JU334404.1 | - | 1297 | 81 | ||
| JU334405.1 | - | 1477 | 81 | |||
| JU486058.1 | - | 1319 | 81 | |||
| JV046992.1 | - | 1414 | 81 | |||
Set Membership: |
|