Search
Search

PLCD1| Gene Symbol: PLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089399.4 | XP_001089399.1 | 944 | 75 | ||
| XM_015131355.1 | XP_014986841.1 | 415 | 75 | |||
| GenBank mRNA | JU321405.1 | - | 318 | 75 | ||
| JV044000.1 | - | 261 | 75 | |||
| JV619461.1 | - | 330 | 75 | |||
Set Membership: |
|