Search
Search

EXOC5| Gene Symbol: EXOC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261248.2 | NP_001248177.1 | 261 | 82 | ||
| XM_028850247.1 | XP_028706080.1 | 387 | 82 | |||
| GenBank mRNA | JU332561.1 | - | 264 | 82 | ||
| JU474034.1 | - | 151 | 82 | |||
| JV045372.1 | - | 264 | 82 | |||
Set Membership: |
|