Search
Search

NUP50| Gene Symbol: NUP50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265689.2 | NP_001252618.1 | 237 | 129 | ||
| GenBank mRNA | JU332594.1 | - | 352 | 129 | ||
| JU474062.1 | - | 376 | 129 | |||
| JV045976.1 | - | 250 | 129 | |||
| JV667328.1 | - | 237 | 129 | |||
Set Membership: |
|