Search
Search

TREH| Gene Symbol: TREH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115876.2 | XP_014971362.2 | 588 | 90 | ||
| XM_028834047.1 | XP_028689880.1 | 588 | 90 | |||
| XM_028834048.1 | XP_028689881.1 | 591 | 90 | |||
| XM_028834049.1 | XP_028689882.1 | 590 | 90 | |||
Set Membership: |
|