Search
Search

TRIM29| Gene Symbol: TRIM29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258046.1 | NP_001244975.1 | 1531 | 133 | ||
| XM_028832635.1 | XP_028688468.1 | 819 | 133 | |||
| GenBank mRNA | JU474992.1 | - | 1531 | 133 | ||
Set Membership: |
|