Search
Search

CBLC| Gene Symbol: CBLC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109370.4 | XP_001109370.1 | 415 | 82 | ||
| XM_015124236.2 | XP_014979722.2 | 416 | 82 | |||
| XM_015124237.2 | XP_014979723.2 | 420 | 82 | |||
| XM_015124238.2 | XP_014979724.1 | 417 | 82 | |||
| XM_028839113.1 | XP_028694946.1 | 418 | 82 | |||
Set Membership: |
|