Search
Search

WSCD1| Gene Symbol: WSCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257765.1 | NP_001244694.1 | 1044 | 93 | ||
| XM_015118307.2 | XP_014973793.2 | 1758 | 93 | |||
| XM_015118308.2 | XP_014973794.2 | 1115 | 93 | |||
| XM_015118309.2 | XP_014973795.2 | 1126 | 93 | |||
| XM_028835628.1 | XP_028691461.1 | 1797 | 93 | |||
| XM_028835629.1 | XP_028691462.1 | 2396 | 93 | |||
| XM_028835630.1 | XP_028691463.1 | 1089 | 93 | |||
| XM_028835631.1 | XP_028691464.1 | 2375 | 93 | |||
| GenBank mRNA | JU333634.1 | - | 1044 | 93 | ||
| JU474889.1 | - | 1039 | 93 | |||
| JV576210.1 | - | 93 | ||||
| JV677963.1 | - | 1051 | 93 | |||