Search
Search

CHRDL2| Gene Symbol: CHRDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115201.2 | XP_014970687.2 | 1976 | 71 | ||
| XM_015115202.2 | XP_014970688.2 | 1918 | 71 | |||
| GenBank mRNA | JU341494.1 | - | 134 | 71 | ||
Set Membership: |
|