Search
Search

GLOD4| Gene Symbol: GLOD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265952.1 | NP_001252881.1 | 187 | 82 | ||
| GenBank mRNA | JU322522.1 | - | 155 | 82 | ||
| JU472652.1 | - | 182 | 82 | |||
| JV044826.1 | - | 187 | 82 | |||
| JV704222.1 | - | 195 | 82 | |||