Search
Search

NOL7| Gene Symbol: NOL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261675.1 | NP_001248604.1 | 638 | 124 | ||
| XM_015135551.2 | XP_014991037.1 | 829 | 124 | |||
| GenBank mRNA | JU334882.1 | - | 645 | 124 | ||
| JV047299.1 | - | 638 | 124 | |||
Set Membership: |
|