Search
Search

STX17| Gene Symbol: STX17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194798.2 | NP_001181727.1 | 839 | 64 | ||
| XM_001112531.4 | XP_001112531.1 | 721 | 64 | |||
| XM_028834381.1 | XP_028690214.1 | 672 | 64 | |||
| GenBank mRNA | JU335200.1 | - | 675 | 64 | ||
| JU472403.1 | - | 665 | 64 | |||
Set Membership: |
|