Search
Search

PNPO| Gene Symbol: PNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265856.1 | NP_001252785.1 | 652 | 93 | ||
| GenBank mRNA | JU335247.1 | - | 651 | 93 | ||
| JU476218.1 | - | 618 | 93 | |||
| JV047512.1 | - | 652 | 93 | |||
Set Membership: |
|