Search
Search

ABCB9| Gene Symbol: ABCB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261296.1 | NP_001248225.1 | 1273 | 65 | ||
| XM_028828903.1 | XP_028684736.1 | 1518 | 65 | |||
| XM_028828904.1 | XP_028684737.1 | 1656 | 65 | |||
| XM_028828905.1 | XP_028684738.1 | 1389 | 65 | |||
| XM_028828906.1 | XP_028684739.1 | 1518 | 65 | |||
| XM_028828907.1 | XP_028684740.1 | 1518 | 65 | |||
| GenBank mRNA | JU333692.1 | - | 1273 | 65 | ||
| JV056440.1 | - | 314 | 65 | |||
Set Membership: |
|