Search
Search

UNC79| Gene Symbol: UNC79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194012.2 | NP_001180941.1 | 3020 | 154 | ||
| XM_015144317.2 | XP_014999803.1 | 5633 | 154 | |||
| XM_015144318.2 | XP_014999804.1 | 5631 | 154 | |||
| XM_015144319.2 | XP_014999805.1 | 5633 | 154 | |||
| XM_015144320.2 | XP_014999806.1 | 3018 | 154 | |||
| XM_015144321.2 | XP_014999807.1 | 5633 | 154 | |||
| XM_015144322.2 | XP_014999808.1 | 5633 | 154 | |||
| XM_015144323.2 | XP_014999809.1 | 5633 | 154 | |||
| XM_015144324.2 | XP_014999810.1 | 5474 | 154 | |||
| XM_015144325.2 | XP_014999811.1 | 5447 | 154 | |||
| XM_015144326.2 | XP_014999812.1 | 5633 | 154 | |||
| XM_015144327.2 | XP_014999813.1 | 5412 | 154 | |||
| XM_015144328.2 | XP_014999814.1 | 2948 | 154 | |||
| XM_015144329.2 | XP_014999815.1 | 5633 | 154 | |||
| XM_015144330.2 | XP_014999816.1 | 5634 | 154 | |||
| XM_028850252.1 | XP_028706085.1 | 5492 | 154 | |||
| XM_028850253.1 | XP_028706086.1 | 3018 | 154 | |||
| GenBank mRNA | JV719770.1 | - | 2700 | 154 | ||