Search
Search

KMT2C| Gene Symbol: KMT2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135341.2 | XP_014990827.2 | 2703 | 105 | ||
| XM_015135342.2 | XP_014990828.2 | 2094 | 105 | |||
| XM_028846497.1 | XP_028702330.1 | 2958 | 105 | |||
| XM_028846498.1 | XP_028702331.1 | 2900 | 105 | |||
| XM_028846499.1 | XP_028702332.1 | 2900 | 105 | |||
| XM_028846500.1 | XP_028702333.1 | 2900 | 105 | |||
| XM_028846501.1 | XP_028702334.1 | 2900 | 105 | |||
| XM_028846502.1 | XP_028702335.1 | 2867 | 105 | |||
| XM_028846503.1 | XP_028702336.1 | 2900 | 105 | |||
| XM_028846504.1 | XP_028702337.1 | 3185 | 105 | |||
| XM_028846505.1 | XP_028702338.1 | 2869 | 105 | |||
| XM_028846506.1 | XP_028702339.1 | 2619 | 105 | |||
| XM_028846507.1 | XP_028702340.1 | 3092 | 105 | |||
Set Membership: |
|