Search
Search

EGLN1| Gene Symbol: EGLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104870.4 | XP_001104870.1 | 4057 | 66 | ||
| XM_015126202.2 | XP_014981688.1 | 1908 | 66 | |||
| XM_028849705.1 | XP_028705538.1 | 1908 | 66 | |||
| GenBank mRNA | JU326539.1 | - | 518 | 66 | ||
| JU326540.1 | - | 518 | 66 | |||
| JV636233.1 | - | 894 | 66 | |||
| JV717360.1 | - | 530 | 66 | |||
Set Membership: |
|