Search
Search

NOL12| Gene Symbol: NOL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193831.3 | NP_001180760.2 | 244 | 94 | ||
| GenBank mRNA | JU336358.1 | - | 244 | 94 | ||
| JU477177.1 | - | 234 | 94 | |||
| JV048232.1 | - | 212 | 94 | |||
| JV661245.1 | - | 218 | 94 | |||