Search
Search

SCRN3| Gene Symbol: SCRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089716.4 | XP_001089716.1 | 849 | 108 | ||
| XM_015110390.2 | XP_014965876.1 | 671 | 108 | |||
| GenBank mRNA | JV723395.1 | - | 839 | 108 | ||
| JV723396.1 | - | 839 | 108 | |||
Set Membership: |
|