Search
Search

SRD5A3| Gene Symbol: SRD5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257949.1 | NP_001244878.1 | 616 | 97 | ||
| XM_015138649.2 | XP_014994135.1 | 775 | 97 | |||
| GenBank mRNA | JU322788.1 | - | 616 | 97 | ||