Search
Search

PROSER1| Gene Symbol: PROSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087439.4 | XP_001087439.2 | 1329 | 99 | ||
| XM_015120959.2 | XP_014976445.2 | 661 | 99 | |||
| GenBank mRNA | JU477301.1 | - | 1317 | 99 | ||
| JU477302.1 | - | 1317 | 99 | |||
| JV048305.1 | - | 654 | 99 | |||
| JV724005.1 | - | 979 | 99 | |||