Search
Search

CCDC68| Gene Symbol: CCDC68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083248.4 | XP_001083248.2 | 956 | 77 | ||
| XM_002800945.3 | XP_002800991.2 | 2259 | 77 | |||
| XM_015122027.2 | XP_014977513.2 | 2263 | 77 | |||
| XM_015122029.2 | XP_014977515.2 | 956 | 77 | |||
| XM_015122030.2 | XP_014977516.2 | 917 | 77 | |||
Set Membership: |
|