Search
Search

PRXL2B| Gene Symbol: PRXL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801736.3 | XP_002801782.2 | 687 | 128 | ||
| XM_002801737.3 | XP_002801783.3 | 543 | 128 | |||
| GenBank mRNA | JU326104.1 | - | 135 | 128 | ||
| JV601946.1 | - | 391 | 128 | |||
Set Membership: |
|