Search
Search

XXYLT1| Gene Symbol: XXYLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194076.3 | NP_001181005.1 | 752 | 64 | ||
| GenBank mRNA | JU327309.1 | - | 288 | 64 | ||
| JV643405.1 | - | 373 | 64 | |||
Set Membership: |
|