Search
Search

EPHX4| Gene Symbol: EPHX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098374.4 | XP_001098374.1 | 594 | 139 | ||
| XM_028831409.1 | XP_028687242.1 | 362 | 139 | |||
| GenBank mRNA | JV463266.1 | - | 312 | 139 | ||
Set Membership: |
|