Search
Search

NDUFAF2| Gene Symbol: NDUFAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266244.1 | NP_001253173.1 | 256 | 150 | ||
| GenBank mRNA | JU323411.1 | - | 315 | 150 | ||
| JV045666.1 | - | 256 | 150 | |||
| JV598251.1 | - | 364 | 150 | |||
Set Membership: |
|