Search
Search

CCDC50| Gene Symbol: CCDC50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132506.2 | XP_014987992.1 | 287 | 80 | ||
| XM_015132507.2 | XP_014987993.1 | 855 | 80 | |||
| XM_015132508.2 | XP_014987994.1 | 287 | 80 | |||
| XM_015132509.2 | XP_014987995.1 | 855 | 80 | |||
| GenBank mRNA | JU323299.1 | - | 557 | 80 | ||
| JU333115.1 | - | 557 | 80 | |||
| JU472327.1 | - | 482 | 80 | |||
| JV046277.1 | - | 403 | 80 | |||
Set Membership: |
|