Search
Search

FAAH2| Gene Symbol: FAAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095907.4 | XP_001095907.1 | 1354 | 135 | ||
| XM_015127556.2 | XP_014983042.1 | 1144 | 135 | |||
| XM_015127557.2 | XP_014983043.1 | 1479 | 135 | |||
Set Membership: |
|