Search
Search

UBR1| Gene Symbol: UBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142046.2 | XP_014997532.1 | 1403 | 124 | ||
| XM_015142047.2 | XP_014997533.1 | 968 | 124 | |||
| XM_028850794.1 | XP_028706627.1 | 1228 | 124 | |||
| GenBank mRNA | JU321000.1 | - | 1344 | 124 | ||
| JU474596.1 | - | 1349 | 124 | |||
Set Membership: |
|