Search
Search

MICU3| Gene Symbol: MICU3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028852366.1 | XP_028708199.1 | 1122 | 133 | ||
| XM_028852367.1 | XP_028708200.1 | 1083 | 133 | |||
| XM_028852368.1 | XP_028708201.1 | 1074 | 133 | |||
| XM_028852369.1 | XP_028708202.1 | 1026 | 133 | |||
| XM_028852370.1 | XP_028708203.1 | 993 | 133 | |||
| GenBank mRNA | JU326223.1 | - | 855 | 133 | ||
| JV614103.1 | - | 1029 | 133 | |||
Set Membership: |
|