Search
Search

SLC16A9| Gene Symbol: SLC16A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096810.4 | XP_001096810.2 | 1805 | 104 | ||
| XM_015147579.2 | XP_015003065.2 | 2210 | 104 | |||
| XM_028826616.1 | XP_028682449.1 | 2084 | 104 | |||
| XM_028826617.1 | XP_028682450.1 | 2135 | 104 | |||
| XM_028826618.1 | XP_028682451.1 | 2057 | 104 | |||
| XM_028826619.1 | XP_028682452.1 | 2031 | 104 | |||
Set Membership: |
|